Learn More
N-Boc-O-ethyl-L-serine, 97%
Product Code. p-7035076
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
Product Code. 15499517
missing translation for 'mfr': Thermo Scientific Alfa Aesar H52781.MD
Request Bulk or Custom Format
CAS: 104839-00-1 | C10H19NO5 | 233.264 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104839-00-1
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.264
MDL Number MFCD06797558
InChI Key KNBDVSSESAKBJM-ZETCQYMHSA-N
Synonym boc-s-2-amino-3-ethoxypropionic acid, s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, 2s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, boco-etser, serine, n-1,1-dimethylethoxy carbonyl-o-ethyl, n-tert-butoxycarbonyl-o-ethylserine, n-boc-2-amino-3-ethoxy-propionic acid, s-n-boc-2-amino-3-ethoxypropionic acid, s-2-tertbutoxycarbonylamino-3-ethoxypropionic acid, n-1,1-dimethylethyl oxy carbonyl-o-ethylserine
PubChem CID 15058451
IUPAC Name (2S)-3-ethoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CCOCC(C(=O)O)NC(=O)OC(C)(C)C
Specifications
Quantity 250 mg
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N-Boc-O-ethyl-L-serine