Learn More
N-Boc-L-threonine methyl ester, 95%
Product Code. p-7053835
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15428108
Brand: Thermo Scientific Alfa Aesar H61545.06
Request Bulk or Custom Format
CAS: 79479-07-5 | C10H19NO5 | 233.26 g/mol
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature 2 - 4°C. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79479-07-5
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD00270479
InChI Key MZMWAPNVRMDIPS-RQJHMYQMSA-N
Synonym boc-thr-ome, 2s,3r-methyl 2-tert-butoxycarbonyl amino-3-hydroxybutanoate, boc-l-threonine methyl ester, n-boc-l-threonine methyl ester, n-tert-butoxycarbonyl-l-threonine methyl ester, n-tert-butoxycarbonyl threonine methyl ester, methyl 2s,3r-2-tert-butoxycarbonyl amino-3-hydroxybutanoate, methyl n-tert-butoxycarbonyl-l-threoninate, n-t-butoxycarbonyl-l-threonine methyl ester
PubChem CID 11368286
IUPAC Name methyl (2S,3R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O
Specifications
Boiling Point 206°C
Color Yellow
Refractive Index 1.45
Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 233.26
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material N-Boc-L-threonine methyl ester