missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-prolinol, 98+%, Thermo Scientific Chemicals

Catalog Number 11302337
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
This item is not returnable. View return policy
69610-40-8
C10H19NO3
201.266
MFCD00066232
BFFLLBPMZCIGRM-QMMMGPOBSA-N
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
643448
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC1CO
This item is not returnable. View return policy
69610-40-8
C10H19NO3
201.266
MFCD00066232
BFFLLBPMZCIGRM-QMMMGPOBSA-N
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
643448
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC1CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 59°C to 62°C
Quantity 250 mg
Beilstein 3542667
Optical Rotation −54° (c=5 in Methanol)
Formula Weight 201.27
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-prolinol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'