Learn More
N-Boc-DL-beta-proline ethyl ester, 97%, Thermo Scientific™
Catalog Number 15403908
Click to view available options
Quantity:
250 mg
1 g
170844-49-2
C12H21NO4
243.30
MFCD03791256
PKDQVUYUWUHNMG-UHFFFAOYNA-N
ethyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-ethyl ester, ethyl 1-boc-beta-prolinate, ethyl 1-boc-3-pyrrolidine-carboxylate, n-boc-dl-beta-proline ethyl ester, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, 1-tert-butyl 3-ethylpyrrolidine-1,3-dicarboxylate, pubchem14290, tert-butyl ethyl pyrrolidine-1,3-dicarboxylate
4192972
1-O-tert-butyl 3-O-ethyl pyrrolidine-1,3-dicarboxylate
CCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C
170844-49-2
C12H21NO4
243.30
MFCD03791256
PKDQVUYUWUHNMG-UHFFFAOYNA-N
ethyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-ethyl ester, ethyl 1-boc-beta-prolinate, ethyl 1-boc-3-pyrrolidine-carboxylate, n-boc-dl-beta-proline ethyl ester, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, 1-tert-butyl 3-ethylpyrrolidine-1,3-dicarboxylate, pubchem14290, tert-butyl ethyl pyrrolidine-1,3-dicarboxylate
4192972
1-O-tert-butyl 3-O-ethyl pyrrolidine-1,3-dicarboxylate
CCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 243.3
Percent Purity 97%
Chemical Name or Material N-Boc-DL-beta-proline ethyl ester