Learn More
N-Boc-3-dimethylamino-L-alanine, 97%
Product Code. 15499567
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15499567
Brand: Thermo Scientific Alfa Aesar H52810.03
Request Bulk or Custom Format
CAS: 94778-71-9 | C10H20N2O4 | 232.28 g/mol
N-Boc-3-dimethylamino-L-alanine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-3-dimethylamino-L-alanine is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 94778-71-9
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD02682423
InChI Key VCDQZVYJKDSORW-ZETCQYMHSA-N
Synonym boc-aza-l-leucine, n-tert-butoxycarbonyl-3-dimethylamino-l-alanine, 2s-2-tert-butoxycarbonyl amino-3-dimethylamino propanoic acid, boc-, ?-n,n-dimethylamino-l-ala, boc-beta-n,n-dimethylamino-l-ala, na-boc-s-2-amino-3-dimethylamino propionic acid, n-alpha-boc-s-2-amino-3-dimethylamino propionic acid, s-2-tert-butoxycarbonylamino-3-dimethylamino-propionic acid, l-alanine,3-dimethylamino-n-1,1-dimethylethoxy carbonyl, 2s-2-tert-butoxy carbonyl amino-3-dimethylamino propanoic acid
PubChem CID 17998956
IUPAC Name (2S)-3-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CN(C)C)C(=O)O
Specifications
Quantity 1 g
Formula Weight 232.28
Percent Purity 97%
Chemical Name or Material N-Boc-3-dimethylamino-L-alanine