missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-chloro-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog Number p-7034334
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
114873-03-9
C14H18ClNO4
299.751
MFCD00672515
RCZHBTHQISEPPP-NSHDSACASA-N
boc-3-chloro-l-phenylalanine, boc-phe 3-cl-oh, boc-l-3-chlorophenylalanine, s-n-boc-3-chlorophenylalanine, boc-l-phe 3-cl-oh, boc-l-3-chlorophe, boc-d-3-chlorophenylalanine, boc-l-3-chloro-phe-oh, s-2-tert-butoxycarbonyl amino-3-3-chlorophenyl propanoic acid, n-boc-3-chloro-l-phenylalanine
2761465
(2S)-3-(3-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Cl)C(=O)O
This item is not returnable. View return policy
114873-03-9
C14H18ClNO4
299.751
MFCD00672515
RCZHBTHQISEPPP-NSHDSACASA-N
boc-3-chloro-l-phenylalanine, boc-phe 3-cl-oh, boc-l-3-chlorophenylalanine, s-n-boc-3-chlorophenylalanine, boc-l-phe 3-cl-oh, boc-l-3-chlorophe, boc-d-3-chlorophenylalanine, boc-l-3-chloro-phe-oh, s-2-tert-butoxycarbonyl amino-3-3-chlorophenyl propanoic acid, n-boc-3-chloro-l-phenylalanine
2761465
(2S)-3-(3-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Cl)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 299.75
Percent Purity 95%
Chemical Name or Material N-Boc-3-chloro-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'