missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-cyclohexyl-D-glycine, 95%, Thermo Scientific™

Product Code. p-7055162
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15400489 1 g 1g
15410489 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15400489

Brand: Thermo Scientific Alfa Aesar H63673.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70491-05-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD00133629
InChI Key QSUXZIPXYDQFCX-SNVBAGLBSA-N
Synonym boc-d-chg-oh, boc-d-cyclohexylglycine, r-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-alpha-cyclohexyl-d-glycine, n-boc-2-cyclohexyl-d-glycine, 2r-n-tert-butoxycarbonyl-2-cyclohexylglycine, r-tert-butoxycarbonylamino-cyclohexyl-acetic acid, r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-2-tert-butoxycarbonylamino-2-cyclohexyl-acetic acid
PubChem CID 7014890
IUPAC Name (2R)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O

Specifications

Beilstein 7689661
Quantity 1 g
Formula Weight 257.33
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-2-cyclohexyl-D-glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.