Learn More
N-BOC-1-Aminocyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-4229094
Click to view available options
Quantity:
1 g
5 g
120728-10-1
C10H16NO4
214.24
MFCD02682623
ROVVUKFHORPDSM-UHFFFAOYSA-M
n-boc-1-aminocyclobutanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 1-boc-amino cyclobutanecarboxylic acid, boc-cyclovaline, boc-acbc-oh, 1-n-boc-amino-cyclobutane carboxylic acid, 1-tert-butoxycarbonylamino cyclobutanecarboxylic acid, boc-1-aminocyclobutane-1-carboxylic acid, n-boc-1-aminocyclobutane carboxylic acid, boc-1-aminocyclobutanecarboxylic acid
1512646
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCC1)C([O-])=O
120728-10-1
C10H16NO4
214.24
MFCD02682623
ROVVUKFHORPDSM-UHFFFAOYSA-M
n-boc-1-aminocyclobutanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 1-boc-amino cyclobutanecarboxylic acid, boc-cyclovaline, boc-acbc-oh, 1-n-boc-amino-cyclobutane carboxylic acid, 1-tert-butoxycarbonylamino cyclobutanecarboxylic acid, boc-1-aminocyclobutane-1-carboxylic acid, n-boc-1-aminocyclobutane carboxylic acid, boc-1-aminocyclobutanecarboxylic acid
1512646
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCC1)C([O-])=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 129°C to 133°C
CAS Min % 96.0
CAS Max % 100.0
Color White to Brown
Assay Percent Range 97%
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 215.25
Percent Purity 97%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-BOC-1-Aminocyclobutanecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-1-Aminocyclobutanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.