Learn More
N-Benzylhydroxylamine hydrochloride, 98%, BLD Pharm™
Product Code. 30968262
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968262 5 g 5g
30967382 100 g 100g
30957226 1 kg 1kg
30958591 500 g 500g
30957210 25 g 25g
30955606 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30968262 Supplier BLD Pharm Supplier No. BD1434905g
 Restricted Item
Estimated Shipment: 27-11-2026
Spezifikation
CAS 29601-98-7
Molecular Formula C7H10ClNO
MDL Number MFCD00043377
Quantity 5 g
SMILES ONCC1=CC=CC=C1.[H]Cl
Molecular Weight (g/mol) 159.61
Percent Purity 98%
Chemical Name or Material N-Benzylhydroxylamine hydrochloride