missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylaminoacetaldehyde diethyl acetal, 96%, Thermo Scientific Chemicals

Catalog Number 11451488
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
61190-10-1
C13H21NO2
223.316
MFCD00051599
SXFVQTYQHWRYOS-UHFFFAOYSA-N
n-benzylaminoacetaldehyde diethyl acetal, benzyl 2,2-diethoxyethyl amine, n-benzyl-n-2,2-diethoxyethyl amine, 2,2-diethoxyethyl benzylamine, acmc-20ak6r, benzylaminoacetaldehyde diethylacetal, benzyl-2,2-diethoxy-ethyl-amine, benzylamino-acetaldehyde diethyl acetal, n-benzyl 2-aminoacetaldehyde diethyl acetal
521961
N-benzyl-2,2-diethoxyethanamine
CCOC(CNCC1=CC=CC=C1)OCC
This item is not returnable. View return policy
61190-10-1
C13H21NO2
223.316
MFCD00051599
SXFVQTYQHWRYOS-UHFFFAOYSA-N
n-benzylaminoacetaldehyde diethyl acetal, benzyl 2,2-diethoxyethyl amine, n-benzyl-n-2,2-diethoxyethyl amine, 2,2-diethoxyethyl benzylamine, acmc-20ak6r, benzylaminoacetaldehyde diethylacetal, benzyl-2,2-diethoxy-ethyl-amine, benzylamino-acetaldehyde diethyl acetal, n-benzyl 2-aminoacetaldehyde diethyl acetal
521961
N-benzyl-2,2-diethoxyethanamine
CCOC(CNCC1=CC=CC=C1)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 115°C to 117°C (1 mmHg)
Refractive Index 1.489
Quantity 1 g
Beilstein 2694610
Formula Weight 223.32
Percent Purity 96%
Chemical Name or Material N-Benzylaminoacetaldehyde diethyl acetal

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'