Learn More
N-Benzyl Salbutamon Hydrochloride, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | 2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride |
| CAS | 24085-08-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 H25 N O3 . Cl H |
| Quantity | 25 mg |
| Synonym | 2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride, Salbutamol Sulphate Imp. G (EP) as Hydrochloride |
| InChI Formula | InChI=1S/C20H25NO3.ClH/c1-20(2,3)21(12-15-7-5-4-6-8-15)13-19(24)16-9-10-18(23)17(11-16)14-22;/h4-11,22-23H,12-14H2,1-3H3;1H |
| SMILES | Cl.CC(C)(C)N(CC(=O)c1ccc(O)c(CO)c1)Cc2ccccc2 |
| IUPAC Name | 2-[benzyl(tert-butyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone;hydrochloride |
| Molecular Weight (g/mol) | 363.88 |
| Show More |