missing translation for 'onlineSavingsMsg'
Learn More

Scharlab N-AMYLALKOHOL PS 1L

Brand:  Scharlab AL01271000


N-AMYLALKOHOL PS 1L

Product Code. 11319853

  • ONLINE EXCLUSIVE PRICE
    597.45 NOK
    523.11 NOK / 1 L
    Save 74.34 NOK
    12% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Amyl alcohol, synthesis grade

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.