Learn More
Thermo Scientific™ N(alpha)-Ethoxycarbonyl-L-asparagine, 97%
455.00 NOK - 1510.00 NOK
Chemical Identifiers
CAS | 16639-91-1 |
---|---|
Molecular Formula | C7H12N2O5 |
Molecular Weight (g/mol) | 204.182 |
MDL Number | MFCD00082761 |
InChI Key | YORYXPTWPIKBDE-BYPYZUCNSA-N |
Synonym | eoc-l-asparagine, l-asparagine, n2-ethoxycarbonyl, n alpha-ethoxycarbonyl-l-asparagine, s-4-amino-2-ethoxycarbonyl amino-4-oxobutanoic acid, n-carbethoxy-l-asparagine, n∼2∼-ethoxycarbonyl-l-asparagine, 2s-3-carbamoyl-2-ethoxycarbonyl amino propanoic acid, n a ethoxycarbonyl-l-asparagine, l-asparagine,n2-ethoxycarbonyl, n alpha-carbethoxy-l-asparagine |
PubChem CID | 7021476 |
IUPAC Name | (2S)-4-amino-2-(ethoxycarbonylamino)-4-oxobutanoic acid |
SMILES | CCOC(=O)NC(CC(=O)N)C(=O)O |
Description
Chemical Identifiers
16639-91-1 | |
204.182 | |
YORYXPTWPIKBDE-BYPYZUCNSA-N | |
7021476 | |
CCOC(=O)NC(CC(=O)N)C(=O)O |
C7H12N2O5 | |
MFCD00082761 | |
eoc-l-asparagine, l-asparagine, n2-ethoxycarbonyl, n alpha-ethoxycarbonyl-l-asparagine, s-4-amino-2-ethoxycarbonyl amino-4-oxobutanoic acid, n-carbethoxy-l-asparagine, n∼2∼-ethoxycarbonyl-l-asparagine, 2s-3-carbamoyl-2-ethoxycarbonyl amino propanoic acid, n a ethoxycarbonyl-l-asparagine, l-asparagine,n2-ethoxycarbonyl, n alpha-carbethoxy-l-asparagine | |
(2S)-4-amino-2-(ethoxycarbonylamino)-4-oxobutanoic acid |
Specifications
∽166°C (decomposition) | |
C7H12N2O5 | |
1712104 | |
YORYXPTWPIKBDE-BYPYZUCNSA-N | |
(2S)-4-amino-2-(ethoxycarbonylamino)-4-oxobutanoic acid | |
7021476 | |
97% |
16639-91-1 | |
MFCD00082761 | |
eoc-l-asparagine, l-asparagine, n2-ethoxycarbonyl, n alpha-ethoxycarbonyl-l-asparagine, s-4-amino-2-ethoxycarbonyl amino-4-oxobutanoic acid, n-carbethoxy-l-asparagine, n∼2∼-ethoxycarbonyl-l-asparagine, 2s-3-carbamoyl-2-ethoxycarbonyl amino propanoic acid, n a ethoxycarbonyl-l-asparagine, l-asparagine,n2-ethoxycarbonyl, n alpha-carbethoxy-l-asparagine | |
CCOC(=O)NC(CC(=O)N)C(=O)O | |
204.182 | |
204.18 | |
N(alpha)-Ethoxycarbonyl-L-asparagine |
Safety and Handling
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold