Learn More
Thermo Scientific Chemicals N(alpha)-Boc-L-asparagine, 98+%
Product Code. p-7023001
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11428646
Brand: Thermo Scientific Chemicals A16019.06
Request Bulk or Custom Format
CAS: 7536-55-2 | C9H16N2O5 | 232.236 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7536-55-2
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00038152
InChI Key FYYSQDHBALBGHX-YFKPBYRVSA-N
Synonym boc-asn-oh, boc-l-asparagine, boc-asn, n-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, nalpha-tert-butoxycarbonyl-l-asparagine, boc-asparagine, tert-butoxycarbonyl-l-asparagine, n-tert-butoxycarbonyl asparagine, tert-butyloxycarbonyl-l-asparagine
PubChem CID 82035
ChEBI CHEBI:3146
IUPAC Name (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O
Specifications
Melting Point ∼170°C (decomposition)
Beilstein 1977963
Quantity 5 g
Formula Weight 232.24
Optical Rotation −7.3° (c=2 in DMF)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-L-asparagine