Learn More
Thermo Scientific Chemicals N(alpha)-Boc-D-tryptophan, 97%
Product Code. p-7045042
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11397286
Brand: Thermo Scientific Chemicals L09214.03
Request Bulk or Custom Format
CAS: 5241-64-5 | C16H20N2O4 | 304.35 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5241-64-5
Molecular Formula C16H20N2O4
Molecular Weight (g/mol) 304.35
MDL Number MFCD00037944
InChI Key NFVNYBJCJGKVQK-CYBMUJFWSA-N
Synonym boc-d-trp-oh, n-boc-d-tryptophan, n-tert-butoxy carbonyl-d-tryptophan, n-tert-butoxycarbonyl-d-tryptophan, boc-d-tryptophan, boc-d-trp, nalpha-boc-d-tryptophan, boc-d-tryptophane, nalpha-boc-d-tryptophane, tert-butoxycarbonyl
PubChem CID 111050
IUPAC Name (2R)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O
Specifications
Melting Point ∽138°C (decomposition)
Beilstein 4237334
Quantity 1 g
Formula Weight 304.35
Optical Rotation +21° (c=1 in Acetic Acid)
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-D-tryptophan