missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Benzyloxycarbonyl-L-glutamine, 99%

Product Code. 15453177
Click to view available options
Quantity:
5 g
25 g
100 g
1000 g
Unit Size:
1000g
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15453177

Brand: Thermo Scientific Alfa Aesar A18240.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2650-64-8 | C13H16N2O5 | 280.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2650-64-8
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00008043
InChI Key JIMLDJNLXLMGLX-JTQLQIEISA-N
Synonym z-gln-oh, n-carbobenzyloxy-l-glutamine, cbz-l-glutamine, cbz-gln-oh, n-carbobenzoxy-l-glutamine, benzyloxycarbonyl-l-glutamine, carbobenzoxyglutamine, n-cbz-l-glutamine, cbz-l-gln-oh, carbobenzoxy-l-glutamine
PubChem CID 75855
IUPAC Name (2S)-5-amino-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES NC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 134°C to 137°C
Beilstein 2061271
Quantity 1000 g
Formula Weight 280.28
Optical Rotation −7° (c=2 in Ethanol)
Percent Purity 99%
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-glutamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.