Learn More
Thermo Scientific Chemicals N-alpha-Acetyl-L-glutamine, 99%
Product Code. 11306996
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
11306996 250 g 250g
11396986 50 g 50g
2 options
Product Code. 11306996
Brand: Thermo Scientific Chemicals L06780.30
Request Bulk or Custom Format
CAS: 2490-97-3 | C7H12N2O4 | 188.183 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2490-97-3
Molecular Formula C7H12N2O4
Molecular Weight (g/mol) 188.183
MDL Number MFCD00038159
InChI Key KSMRODHGGIIXDV-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
PubChem CID 182230
ChEBI CHEBI:21553
IUPAC Name (2S)-2-acetamido-5-amino-5-oxopentanoic acid
SMILES CC(=O)NC(CCC(=O)N)C(=O)O
Specifications
Melting Point ∽206°C (decomposition)
Beilstein 1727471
Merck Index 14,25
Quantity 250 g
Formula Weight 188.18
Optical Rotation −12° (c=3 in Water)
Percent Purity 99%
Chemical Name or Material N-alpha-Acetyl-L-glutamine