missing translation for 'onlineSavingsMsg'
Learn More

N-Acryloylsarcosine methyl ester, 97%, stab. with 250ppm 4-methoxyphenol, Thermo Scientific™

Catalog Number 11337916
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
72065-23-7
C7H11NO3
157.17
MFCD00151792
ZCQGVFNHUATAJY-UHFFFAOYSA-N
n-acryloylsarcosine methyl ester, acryloylsarcosine methyl ester, methyl 2-n-methylprop-2-enamido acetate, methyl 2-n-methylacrylamido acetate, n-acryloyl-n-methylglycine methyl ester, methyl 2-n-methylprop-2-enoylamino acetate, methyl 2-methyl prop-2-enoyl amino ethanoate, 2-methyl 1-oxoprop-2-enyl amino acetic acid methyl ester, glycine,n-methyl-n-1-oxo-2-propen-1-yl-, methyl ester
7015777
methyl 2-[methyl(prop-2-enoyl)amino]acetate
COC(=O)CN(C)C(=O)C=C
This item is not returnable. View return policy
72065-23-7
C7H11NO3
157.17
MFCD00151792
ZCQGVFNHUATAJY-UHFFFAOYSA-N
n-acryloylsarcosine methyl ester, acryloylsarcosine methyl ester, methyl 2-n-methylprop-2-enamido acetate, methyl 2-n-methylacrylamido acetate, n-acryloyl-n-methylglycine methyl ester, methyl 2-n-methylprop-2-enoylamino acetate, methyl 2-methyl prop-2-enoyl amino ethanoate, 2-methyl 1-oxoprop-2-enyl amino acetic acid methyl ester, glycine,n-methyl-n-1-oxo-2-propen-1-yl-, methyl ester
7015777
methyl 2-[methyl(prop-2-enoyl)amino]acetate
COC(=O)CN(C)C(=O)C=C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.02
Boiling Point 96°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.477
Quantity 2 g
Beilstein 4964477
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material N-Acryloylsarcosine methyl ester, stabilized with 250ppm 4-methoxyphenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'