Learn More
N-Acetyl-S-(3-hydroxypropyl)-L-cysteine Dicyclohexylamine Salt, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-S-(3-hydroxypropyl)-L-cysteine dicyclohexylammonium salt |
| CAS | 14369-42-7 |
| Molecular Formula | C12 H23 N . C8 H15 N O4 S |
| Synonym | L-Cysteine, N-acetyl-S-(3-hydroxypropyl)-, compd. with N-cyclohexylcyclohexanamine (1:1), Alanine, N-acetyl-3-[(3-hydroxypropyl)thio]-, compd. with dicyclohexylamine (1:1), L- (8 CI), Alanine, N-acetyl-3-[(3-hydroxypropyl)thio]-, compd. with dicyclohexylamine (7 CI), Dicyclohexylamine, compd. with N-acetyl-3-[(3-hydroxypropyl)thio]-L-alanine (1:1), N-Acetyl-S-(3-hydroxypropyl)cysteine dicyclohexylammonium salt, N-Acetyl-S-(3-hydroxypropyl)-L-cysteine dicyclohexylammonium salt |
| InChI Formula | InChI=1 S/C12H23N.C8H15NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-6(11)9-7(8(12)13)5-14-4-2-3-10/h11-13 H,1-10H2;7,10 H,2-5H2,1H3,(H,9,11)(H,12,13)/t;7-/m0.0/s1 |
| SMILES | CC(=O)N[C@@H](CSCCCO)C(=O)O.C1CCC(CC1)NC2CCCCC2 |
| IUPAC Name | (2 R)-2-acetamido-3-(3-hydroxypropylsulfanyl)propanoic acid;N-cyclohexylcyclohexanamine |
| Molecular Weight (g/mol) | 402.59 |
| Quantity | 0.01 g |
| Formula Weight | 402.2552 g/mol |
| Show More |