missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-L-arginine dihydrate, >99%, MP Biomedicals™

Product Code. 11328379 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11328379

Brand: MP Biomedicals 0210007810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 155-84-0
Color White
Assay Percent Range >99%
Molecular Formula C8H16N4O3
MDL Number MFCD00019733
Synonym n-acetyl-l-arginine, n-alpha-l-acetyl-arginine, n2-acetyl-l-arginine, arginine, n2-acetyl, ac-arg-oh.2h2o, unii-tq7dl04cae, s-2-acetamido-5-guanidinopentanoic acid, ac-arg-oh, n-a-acetyl-l-arginine, nalpha-acetyl-l-arginine
InChI Key SNEIUMQYRCDYCH-LURJTMIESA-N
SMILES CC(=O)NC(CCCN=C(N)N)C(=O)O
IUPAC Name (2S)-2-acetamido-5-(diaminomethylideneamino)pentanoic acid
Molecular Weight (g/mol) 216.241
Quantity 10 g
PubChem CID 67427
ChEBI CHEBI:40521
Formula Weight 252.27
Physical Form Powder
Chemical Name or Material N-acetyl-L-arginine dihydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.