Learn More
N-Acetyl-DL-tryptophan, 99%
Product Code. p-7023724
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11478187
Brand: Thermo Scientific Alfa Aesar A17562.14
Request Bulk or Custom Format
CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol
N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.
Solubility
Soluble in 1N Sodium Hydroxide, water, and methanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87-32-1
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00005644
InChI Key DZTHIGRZJZPRDV-UHFFFAOYSA-N
Synonym n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
PubChem CID 2002
ChEBI CHEBI:70976
IUPAC Name 2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
Specifications
Melting Point ∼205°C (decomposition)
Beilstein 89478
Solubility Information Soluble in 1N Sodium Hydroxide,water,and methanol.
Quantity 25 g
Formula Weight 246.27
Percent Purity 99%
Chemical Name or Material N-Acetyl-DL-tryptophan