Learn More
N-Acetyl-d3-5-methoxytryptamine-a,a,b,b-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Melatonin-d7 |
| CAS | 615251-68-8 |
| Molecular Formula | C13 2H7 H9 N2 O2 |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C13H16N2O2/c1-9(16)14-6-5-10-8-15-13-4-3-11(17-2)7-12(10)13/h3-4,7-8,15 H,5-6H2,1-2H3,(H,14,16)/i1D3,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)NC([2 H])([2 H])C([2 H])([2 H])c1c[nH]c2ccc(OC)cc12 |
| IUPAC Name | 2,2,2-trideuterio-N-[1,1,2,2-tetradeuterio-2-(5-methoxy-1 H-indol-3-yl)ethyl]acetamide |
| Molecular Weight (g/mol) | 239.32 |
| Formula Weight | 239.1651 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |