Learn More
Thermo Scientific Chemicals N-Acetyl-D-sphingosine, 98%
Product Code. 15443289
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 15443289
Brand: Thermo Scientific Chemicals J60534.M¬
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells
  • Activates protein phosphatases
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3102-57-6
Molecular Formula C20H39NO3, C20H39NO3
Synonym Acetyl ceramide; C2-Ceramide
Specifications
Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases
Formula Weight 341.53g/mol
Melting Point 77°C to 78°C
Solubility Soluble in ethanol, methanol, chloroform or DMSO at approximately 10mg/ml
Product Type Signal Transduction Reagent
Form Powder
InChI Key BLTCBVOJNNKFKC-QUDYQQOWSA-N
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(C)=O
IUPAC Name N-[(2S,3R,4E)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
Molecular Weight (g/mol) 341.54
Assay 98%