Learn More
Thermo Scientific™ N-Acetyl-Adrenocorticotropic Hormone (1-17), human
865.00 NOK - 1246.00 NOK
Specifications
Content And Storage Store at -20°C
Format Solid
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15430537
SDS View Documents Promotion Details
Thermo Scientific™
J66257.!D
0.5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15440537
SDS View Documents Promotion Details
Thermo Scientific™
J66257.M^
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Store at -20°C
Solid