Learn More
N-Acetyl-3-acetoxy-5-phenylpyrrole, TRC
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-3-acetoxy-5-phenylpyrrole |
| CAS | 100750-39-8 |
| Molecular Formula | C14 H13 N O3 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C14H13NO3/c1-10(16)15-9-13(18-11(2)17)8-14(15)12-6-4-3-5-7-12/h3-9H,1-2H3 |
| SMILES | CC(=O)Oc1cc(c2ccccc2)n(c1)C(=O)C |
| IUPAC Name | (1-acetyl-5-phenylpyrrol-3-yl) acetate |
| Molecular Weight (g/mol) | 243.26 |
| Formula Weight | 243.0895 |