Learn More
N-(6-Methyl-2-pyridyl)thiourea, 97%
Product Code. 15496767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496767 5 g 5g
15486767 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15496767 Supplier Thermo Scientific Alfa Aesar Supplier No. H33884.06
Request Bulk or Custom Format
CAS: 49600-34-2 | C7H9N3S | 167.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 49600-34-2
Molecular Formula C7H9N3S
Molecular Weight (g/mol) 167.23
MDL Number MFCD00462739
InChI Key LEWSYNYKTYXRHP-UHFFFAOYSA-N
Synonym 6-methyl-pyridin-2-yl-thiourea, 6-methylpyridin-2-yl thiourea, n-6-methylpyridin-2-yl thiourea, 1-6-methylpyridin-2-yl thiourea, thiourea, 6-methyl-2-pyridyl, n-6-methyl-2-pyridyl thiourea, n-6-methyl-2-pyridinyl thiourea #, thiourea, n-6-methyl-2-pyridinyl, thiourea,n-6-methyl-2-pyridinyl
PubChem CID 685983
IUPAC Name (6-methylpyridin-2-yl)thiourea
SMILES CC1=CC=CC(NC(N)=S)=N1
Specifications
Melting Point 193°C to 197°C
Quantity 5 g
Formula Weight 167.23
Percent Purity 97%
Chemical Name or Material N-(6-Methyl-2-pyridyl)thiourea