missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Methyl-2-pyridyl)thiourea, 97%, Thermo Scientific™

Product Code. 15417317
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15417317 1 g 1g
15427317 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15417317

Brand: Thermo Scientific Alfa Aesar H34378.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131185-00-7
Molecular Formula C7H9N3S
Molecular Weight (g/mol) 167.23
MDL Number MFCD04117347
InChI Key GEYNPFWKKUCREI-UHFFFAOYSA-N
Synonym 5-methyl-pyridin-2-yl-thiourea, n-5-methylpyridin-2-yl thiourea, 5-methylpyridin-2-yl thiourea, 1-5-methylpyridin-2-yl thiourea, n-5-methyl-2-pyridyl thiourea, 5-methylpyridin-2-yl-thiourea, n-5-methyl-2-pyridinyl thiourea, thiourea, n-5-methyl-2-pyridinyl
PubChem CID 2760060
IUPAC Name (5-methylpyridin-2-yl)thiourea
SMILES CC1=CN=C(NC(N)=S)C=C1

Specifications

Melting Point 199°C to 201°C
Quantity 1 g
Formula Weight 167.23
Percent Purity 97%
Chemical Name or Material N-(5-Methyl-2-pyridyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.