missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Bromo-6-methyl-2-pyridyl)thiourea, 97%, Thermo Scientific™

Catalog Number 15496987
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1823232-26-3
C7H8BrN3S
246.126
MFCD22125033
ICVSYPAXVIRTJK-UHFFFAOYSA-N
n-5-bromo-6-methyl-2-pyridyl thiourea
98000722
(5-bromo-6-methylpyridin-2-yl)thiourea
CC1=C(C=CC(=N1)NC(=S)N)Br
This item is not returnable. View return policy
1823232-26-3
C7H8BrN3S
246.126
MFCD22125033
ICVSYPAXVIRTJK-UHFFFAOYSA-N
n-5-bromo-6-methyl-2-pyridyl thiourea
98000722
(5-bromo-6-methylpyridin-2-yl)thiourea
CC1=C(C=CC(=N1)NC(=S)N)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 238°C to 242°C
Quantity 250 mg
Formula Weight 246.12
Percent Purity 97%
Chemical Name or Material N-(5-Bromo-6-methyl-2-pyridyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'