Learn More
N-[4-Nitro-3-(trifluoromethyl)phenyl]-propanamide, TRC
Product Code. 18033340
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18033340 1 g 1g
18072140 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18033340 Supplier Toronto Research Chemicals Supplier No. TRCN5968501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13312-12-4
Molecular Formula C10 H9 F3 N2 O3
Molecular Weight (g/mol) 262.19
Synonym N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide, Flutamide Imp. C (EP)
IUPAC Name N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
SMILES CCC(=O)Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H9F3N2O3/c1-2-9(16)14-6-3-4-8(15(17)18)7(5-6)10(11,12)13/h3-5H,2H2,1H3,(H,14,16)
Formula Weight 262.0565
Chemical Name or Material N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide