Learn More
N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide, Mikromol™
Product Code. 30459242
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459242 4 x 25 mg -
30052476 25 mg 25mg
18004010 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30459242 Supplier Mikromol Supplier No. MM0161.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide
CAS 13312-12-4
Analyte or Component Names N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide
Shipping Condition Room Temperature
Molecular Formula C10 H9 F3 N2 O3
Certifications/Compliance ISO 17025
Synonym Flutamide Imp. C (EP), N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide, Flutamide Impurity C
InChI Formula InChI=1S/C10H9F3N2O3/c1-2-9(16)14-6-3-4-8(15(17)18)7(5-6)10(11,12)13/h3-5H,2H2,1H3,(H,14,16)
Solution Type Neat
SMILES CCC(=O)Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]
IUPAC Name N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Molecular Weight (g/mol) 262.19
Quantity 4 x 25 mg
Formula Weight 262.0565 g/mol