Learn More
N-(4-Fluorobenzyl)-3-methoxyaniline, 97%, Thermo Scientific™
Product Code. 15493228
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493228 250 mg 250mg
15483228 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15493228 Supplier Thermo Scientific Alfa Aesar Supplier No. H56708.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 356531-63-0
Molecular Formula C14H14FNO
Molecular Weight (g/mol) 231.27
MDL Number MFCD03210777
InChI Key LFWQQAPNOAPZNW-UHFFFAOYSA-N
Synonym n-4-fluorobenzyl-3-methoxyaniline, n-4-fluorophenyl methyl-3-methoxyaniline
PubChem CID 24160879
IUPAC Name N-[(4-fluorophenyl)methyl]-3-methoxyaniline
SMILES COC1=CC=CC(NCC2=CC=C(F)C=C2)=C1
Specifications
Quantity 250 mg
Formula Weight 231.26
Percent Purity 97%
Chemical Name or Material N-(4-Fluorobenzyl)-3-methoxyaniline