Learn More
N-(4-Carboxyphenyl)succinamic acid, 99%, Thermo Scientific Chemicals
Product Code. p-7047080
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11348496 1 g 1g
11358496 5 g 5g
2 options
Product Code. 11348496
Brand: Thermo Scientific Alfa Aesar L17549.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76475-62-2
Molecular Formula C11H11NO5
Molecular Weight (g/mol) 237.21
MDL Number MFCD00020530
InChI Key IOKXKKSBNJCKOY-UHFFFAOYSA-N
Synonym 4-3-carboxypropanamido benzoic acid, 4-3-carboxypropanoyl amino benzoic acid, n-4-carboxyphenyl succinamic acid, 4-3-carboxypropanoylamino benzoic acid, cbkinase1_000202, cbkinase1_012602, cambridge id 5122038, n-4-carboxyphenyl succinamidic acid, 4-2-carboxyethylcarboxamido benzoic acid
PubChem CID 765581
IUPAC Name 4-(3-carboxypropanoylamino)benzoic acid
SMILES OC(=O)CCC(=O)NC1=CC=C(C=C1)C(O)=O
Specifications
Melting Point ∽290°C (decomposition)
Quantity 1 g
Beilstein 3355574
Formula Weight 237.21
Percent Purity 99%
Chemical Name or Material N-(4-Carboxyphenyl)succinamic acid