missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromobutyl)phthalimide, 96%

Product Code. 11451868
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11451868

Brand: Thermo Scientific Alfa Aesar A14517.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5394-18-3 | C12H12BrNO2 | 282.14 g/mol

N-(4-Bromobutyl)phthalimide is used in organic synthesis and the production of pharmaceutical. It can react with 1-phenyl-piperazine to get N-[4-(4-phenyl-piperazin-1-yl)-butyl]-phthalimide. It is a useful synthesis reagent used to synthesize B-cyclodextrin derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Bromobutyl)phthalimide is used in organic synthesis and the production of pharmaceutical. It can react with 1-phenyl-piperazine to get N-[4-(4-phenyl-piperazin-1-yl)-butyl]-phthalimide. It is a useful synthesis reagent used to synthesize B-cyclodextrin derivatives.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store at room temperature. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5394-18-3
Molecular Formula C12H12BrNO2
Molecular Weight (g/mol) 282.14
MDL Number MFCD00005905
InChI Key UXFWTIGUWHJKDD-UHFFFAOYSA-N
Synonym n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide
PubChem CID 93575
IUPAC Name 2-(4-bromobutyl)isoindole-1,3-dione
SMILES BrCCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 77°C to 81°C
Boiling Point 165°C to 170°C (1 mmHg)
Odor Odorless
Quantity 25 g
Beilstein 164119
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 282.14
Percent Purity 98%
Chemical Name or Material N-(4-Bromobutyl)phthalimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.