Learn More
N-[[4-[[(4-Amino[1,1'-biphenyl]-3-yl)amino]carbonyl]phenyl]methyl]carbamic Acid Methyl Ester-d3, TRC
Specifications
Specifications
| Chemical Name or Material | N-[(4-[((4-Amino[1,1'-biphenyl]-3-yl)amino)carbonyl]phenyl)methyl]carbamic Acid Methyl Ester-d3 |
| Molecular Formula | C22 H18 D3 N3 O3 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C22H21N3O3/c1-28-22(27)24-14-15-7-9-17(10-8-15)21(26)25-20-13-18(11-12-19(20)23)16-5-3-2-4-6-16/h2-13H,14,23H2,1H3,(H,24,27)(H,25,26)/i1D3 |
| SMILES | [2H]C([2H])([2H])OC(=O)NCc1ccc(cc1)C(=O)Nc2cc(ccc2N)c3ccccc3 |
| IUPAC Name | trideuteriomethyl N-[[4-[(2-amino-5-phenylphenyl)carbamoyl]phenyl]methyl]carbamate |
| Molecular Weight (g/mol) | 378.439 |
| Formula Weight | 378.177 |