Learn More
N-[[4-[2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy]phenyl]methyl]glycine Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | N-((4-(2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy)phenyl)methyl)glycine Methyl Ester |
| CAS | 331746-65-7 |
| Molecular Formula | C22 H24 N2 O4 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C22H24N2O4/c1-16-20(24-22(28-16)18-6-4-3-5-7-18)12-13-27-19-10-8-17(9-11-19)14-23-15-21(25)26-2/h3-11,23H,12-15H2,1-2H3 |
| SMILES | COC(=O)CNCc1ccc(OCCc2nc(oc2C)c3ccccc3)cc1 |
| Molecular Weight (g/mol) | 380.437 |
| Formula Weight | 380.174 |