missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Chlorophenylsulfonyl)-S-methyl-DL-homocysteine, 96%, Thermo Scientific™

Catalog Number 15478905
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
C11H14ClNO4S2
MFCD06409389
This item is not returnable. View return policy
C11H14ClNO4S2
MFCD06409389

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 103°C
Formula Weight 323.82
Percent Purity 96%
Quantity 250 mg
Chemical Name or Material N-(3-Chlorophenylsulfonyl)-S-methyl-DL-homocysteine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'