Learn More
N-(3-Aminopropyl)cyclohexylamine, 98%, Thermo Scientific™
Product Code. 11347646
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347646 100 g 100g
11337646 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11347646 Supplier Thermo Scientific Alfa Aesar Supplier No. L11250.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3312-60-5
Molecular Formula C9H20N2
Molecular Weight (g/mol) 156.273
MDL Number MFCD00003838
InChI Key ITZPOSYADVYECJ-UHFFFAOYSA-N
Synonym n-3-aminopropyl cyclohexylamine, n1-cyclohexylpropane-1,3-diamine, 1,3-propanediamine, n-cyclohexyl, n-cyclohexyl-1,3-propanediamine, cyclohexyl-1,3-propanediamine, 3-aminopropyl cyclohexylamine, n-cyclohexyl-1,3-diaminopropane, unii-f4xob09fkn, n-cyclohexyl-1,3-propylenediamine, n-cyclohexylpropylene 1,3-diamine
PubChem CID 18718
IUPAC Name N'-cyclohexylpropane-1,3-diamine
SMILES C1CCC(CC1)NCCCN
Specifications
Melting Point -18°C
Density 0.92
Boiling Point 236°C
Flash Point 111°C (232°F)
Refractive Index 1.482
Quantity 100 g
UN Number UN2735
Beilstein 741954
Sensitivity Air Sensitive
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material N-(3-Aminopropyl)cyclohexylamine