Learn More
N-[3-[3-(2-Amino-2-oxoethyl)-1H-indol-1-yl]propyl]-carbamic Acid 1,1-Dimethylethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | N-[3-[3-(2-Amino-2-oxoethyl)-1H-indol-1-yl]propyl]-carbamic Acid 1,1-Dimethylethyl Ester |
| CAS | 1024583-85-4 |
| Molecular Formula | C18H25N3O3 |
| Quantity | 100 mg |
| Synonym | t-Butyl (3-(3-(2-Amino-2-oxoethyl)-1H-indol-1-yl)propyl)carbamate |
| InChI Formula | InChI=1S/C18H25N3O3/c1-18(2,3)24-17(23)20-9-6-10-21-12-13(11-16(19)22)14-7-4-5-8-15(14)21/h4-5,7-8,12H,6,9-11H2,1-3H3,(H2,19,22)(H,20,23) |
| SMILES | CC(C)(C)OC(=O)NCCCn1cc(CC(=O)N)c2ccccc12 |
| IUPAC Name | tert-butyl N-[3-[3-(2-amino-2-oxoethyl)indol-1-yl]propyl]carbamate |
| Molecular Weight (g/mol) | 331.41 |