Learn More
Macherey-Nagel™ N 20 Magnetic Crimp Caps
Product Code. 12440490 Shop All Macherey-Nagel Products
Click to view available options
Septum:
Butyl Red/PTFE Gray
Butyl/PTFE
Silicone/PTFE
Color (Seal):
Dark Gray/Gray
Light Gray/Dark Gray
Transparent Blue/Clear
Unit Size:
Pack of 100
Product Code. 12440490
Brand: Macherey-Nagel™ 702928
Only null left
Specifications
Color (Seal) Light Gray/Dark Gray
Diameter (Metric) Opening 8 mm
Color Silver
Material Butyl Rubber, PTFE
Septum Butyl/PTFE
Thickness (Metric) 3 mm