Learn More
N-(2-Chloropropyl)morpholine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N-(2-Chloropropyl)morpholine Hydrochloride |
| CAS | 100859-99-2 |
| Molecular Formula | C7H15Cl2NO |
| Quantity | 100 mg |
| Synonym | 4-(2-Chloropropyl)morpholine Hydrochloride |
| InChI Formula | InChI=1S/C7H14ClNO.ClH/c1-7(8)6-9-2-4-10-5-3-9;/h7H,2-6H2,1H3;1H |
| SMILES | Cl.CC(Cl)CN1CCOCC1 |
| IUPAC Name | 4-(2-chloropropyl)morpholine;hydrochloride |
| Molecular Weight (g/mol) | 200.11 |
| Formula Weight | 199.0531 |