missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromo-4-fluorophenyl)-2-methoxybenzamide, 97%, Thermo Scientific™

Product Code. 15484986
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484986 250 mg 250mg
15455658 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484986 Supplier Thermo Scientific Alfa Aesar Supplier No. H59368.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1021439-52-0
Molecular Formula C14H11BrFNO2
Molecular Weight (g/mol) 324.149
MDL Number MFCD12559362
InChI Key CVEPLYNHBLSXCV-UHFFFAOYSA-N
Synonym n-2-bromo-4-fluorophenyl-2-methoxybenzamide
PubChem CID 28938569
IUPAC Name N-(2-bromo-4-fluorophenyl)-2-methoxybenzamide
SMILES COC1=CC=CC=C1C(=O)NC2=C(C=C(C=C2)F)Br

Specifications

Quantity 250 mg
Formula Weight 324.15
Percent Purity 97%
Chemical Name or Material N-(2-Bromo-4-fluorophenyl)-2-methoxybenzamide
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.