Learn More
N-[(1S)-1-(Hydroxymethyl)-3-phenylpropyl]carbamic Acid-d2 Phenyl Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | N-[(1S)-1-(Hydroxymethyl)-3-phenylpropyl]carbamic Acid-d2 Phenyl Methyl Ester |
| Molecular Formula | C18 D2 H19 N O3 |
| Quantity | 250 mg |
| Synonym | [(1S)-1-(Hydroxymethyl)-3-phenylpropyl]carbamic Acid-d2 Phenylmethyl Ester, Benzyl ((S)-1-Hydroxymethyl-3-phenylpropyl)carbamate-d2 |
| InChI Formula | InChI=1S/C18H21NO3/c20-13-17(12-11-15-7-3-1-4-8-15)19-18(21)22-14-16-9-5-2-6-10-16/h1-10,17,20H,11-14H2,(H,19,21)/t17-/m0/s1/i13D2 |
| SMILES | [2H]C([2H])(O)[C@H](CCc1ccccc1)NC(=O)OCc2ccccc2 |
| IUPAC Name | benzyl N-[(2S)-1,1-dideuterio-1-hydroxy-4-phenylbutan-2-yl]carbamate |
| Molecular Weight (g/mol) | 301.377 |
| Formula Weight | 301.165 |