Specifications
| Color | Silver |
| Material | Aluminum, Butyl Rubber, PTFE |
| Septum | Butyl dark gray / PTFE gray(only centrically laminated with PTFE, typically called Pharma-Fix) |
| Quantity | 100 Pk. |
| Thickness (Metric) | 2 mm |
| Product Type | Crimp Cap |