Learn More
Thermo Scientific™ mySPIN™ 6 Mini Centrifuge
Product Code. 17254243 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
12 V Adapter
Benchtop Mini Centrifuge
Transport Box with Inserts
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17254243 12 V Adapter Each
15334204 Benchtop Mini Centrifuge Each
17244243 Transport Box with Inserts Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17254243 Supplier Thermo Scientific™ Supplier No. 75004087
  • ONLINE EXCLUSIVE PRICE
    690.00 NOK
    608.67 NOK / Pack of 1
    Save 81.33 NOK
    11% Off
 Restricted Item
In Stock
Our most compact centrifuge, the mySPIN 6 mini centrifuge, is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microcentrifuge tubes.
Bring efficiency and user comfort to your lab stations with the Thermo Scientific mySPIN 6 mini centrifuge. Ergonomic design, tool-free rotors, and quick-spin options make it fast and easy to use. Ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2 mL) for customized use.
Enjoy convenience at your personal workstation with our most compact centrifuges, featuring:
  • Small footprint for efficient use of lab space
  • Easy operation and palm-shaped lid for left- or right-handed users
  • Quick-spin feature
  • Quiet operation, low vibration
  • Safety features including unit will not operate without lid in place for safety
  • Includes 2 rotors to accommodate 0.2, 0.5, 1.5, 2.0 mL microcentrifuge tubes and PCR strip tubes
  • Tool-free, quick rotor exchange
TRUSTED_SUSTAINABILITY
Specifications
Product Line mySpin 6
Voltage 12 V
Product Type 12 V Adapter
Does the mySPIN 6 Mini Centrifuge come with a rotor?
The mySPIN 6 Mini Centrifuge includes 2 rotors and adapters to accommodate 0.2, 0.5, 1.5, 2.0 mL microcentrifuge tubes and PCR strip tubes.