Learn More
Myozenin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11450-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Proteintech's Myozenin-2 antibody 11450-1-AP was raised against full-length protein of human origin and is able to detect a band of approximately 34 kDa in Western blotting.
The calcineurin-binding protein Myozenin 2, also designated calsarcin-1, is a member of the calsarcin protein family. Calcineurin is a calcium- and calmodulin-dependent protein phosphatase that is involved in controlling the slow fiber gene expression in skeletal muscle and hypertrophy of cardiac muscle. The calsarcins are sarcomeric proteins that couple calcineurin and muscle activity. In cardiac and skeletal muscle cells, Myozenin 2 binds calcineurin to alpha-actinin at the Z-line of the sarcomere. During embryogenesis, Myozenin 1 and 2 are expressed in developing muscle. The Myozenin 2 gene maps to chromosome 4q and is expressed specifically in adult cardiac and slow-twitch skeletal muscle, while Myozenin 1 is only detected in fast skeletal muscle.Specifications
Myozenin 2 | |
Polyclonal | |
Unconjugated | |
MYOZ2 | |
C4orf5, Calsarcin-1, FATZ related protein 2, MYOZ2, Myozenin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295426, 51778, 59006 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JJW5, Q9NPC6 | |
Myoz2 | |
Myozenin 2 Fusion Protein Ag2008 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |