Learn More
MYOM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22084-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16801635
  • 1611.84 NOK / 20µL
Estimated Shipment: 22-05-2024
Description
The MYOM1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human MYOM1. This antibody recognizes human, mouse, rat antigen. The MYOM1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
MYOM1
Polyclonal
Unconjugated
20 μL
8736
IgG
Western Blot, Immunohistochemistry, ELISA
447 μg/mL
Rabbit
Primary
Human, Rat, Mouse