missing translation for 'onlineSavingsMsg'
Learn More

MYLIP Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15455-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894634

  • 1611.84 NOK / 20µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

The ERM protein family members ezrin, radixin, and moesin are cytoskeletal effector proteins linking actin to membrane-bound proteins at the cell surface. Myosin regulatory light chain interacting protein (MYLIP) is a novel ERM-like protein that interacts with myosin regulatory light chain and inhibits neurite outgrowth.
Specifications

Specifications

MYLIP
Polyclonal
Unconjugated
MYLIP
BZF1, Idol, MIR, MYLIP, PP5242
Rabbit
Antigen Affinity Chromatography
RUO
100760940, 218203, 29116, 306825
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZDI6, Q8BM54, Q8WY64
MYLIP
MYLIP Fusion Protein Ag7720
20 μL
Primary
Human, Hamster, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers