missing translation for 'onlineSavingsMsg'
Learn More

Myeloperoxidase/MPO Antibody (MPO/8631R), PerCP, Novus Biologicals™

Product Code. 18648534 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
18648534 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 18648534

Brand: Novus Biologicals NBP324098PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloperoxidase/MPO
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MPO/8631R
Conjugate PerCP
Formulation PBS
Gene Alias EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Host Species Rabbit
Immunogen Recombinant fragment of human Myeloperoxidase/MPO protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4353
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.