Learn More
Myelin Protein Zero Antibody (MPZ/7390), Novus Biologicals™
Product Code. 18621154 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
Product Code. Quantity unitSize
18621154 100 μg 100µg
18676864 20 μg 20µg
2 options
Product Code. 18621154
Brand: Novus Biologicals NBP323877100ug
Mouse Monoclonal Antibody
Myelin Protein Zero Monoclonal antibody specifically detects Myelin Protein Zero in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Antigen Myelin Protein Zero
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MPZ/7390
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias Charcot-Marie-Tooth neuropathy 1B, CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, HMSNIB, MPP, Myelin peripheral protein, myelin protein P0, myelin protein zeroDSS, P0
Host Species Mouse
Immunogen Recombinant full-length human Myelin Protein Zero protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Oligodendrocyte Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4359
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ