Learn More
MYD88 Mouse anti-Human, Unconjugated, Clone: 2E10D3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67969-1-IG-150UL
 View more versions of this product
Product Code. 17531486
  • 4836.10 NOK / 150µL
Description
The MYD88 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human MYD88. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The MYD88 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
MYD88
Monoclonal
Unconjugated
150 μL
4615
IgG2b
Western Blot, Immunohistochemistry
2E10D3
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit