Learn More
MYD88 Mouse anti-Human, Unconjugated, Clone: 2E10D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67969-1-IG-150UL
Description
The MYD88 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human MYD88. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The MYD88 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| MYD88 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 4615 | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 2E10D3 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit |